2,4-Dibromothiophene ≥90.0% (by GC)

供应商: TCI
Warning

D4146-5G D4146-1G
TCIAD4146-5GEA 0 CNY
TCIAD4146-5G TCIAD4146-1G
2,4-Dibromothiophene ≥90.0% (by GC)
2,4-Dibromothiophene
分子式: C₄H₂Br₂S
沸点: 97°C
溶点: –30 °C
密度: 2,14 g/cm³ (20 °C)
存储条件: Refrigerator
MDL编号: MFCD00043889
CAS号: 3140-92-9
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbc87258cf8bbcf5f871d8ba6e1f3a6eab89e9b2&vid=fbc87258cf8bbcf5f871d8ba6e1f3a6eab89e9b2&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR