3-Chloro-1-fluoro-4-nitrobenzene ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-Chloro-4-fluoro-1-nitrobenzene, 2-Chloro-4-fluoronitrobenzene
C2615-5G
C2615-25G
TCIAC2615-5GEA
0
CNY
TCIAC2615-5G
TCIAC2615-25G
3-Chloro-1-fluoro-4-nitrobenzene ≥98.0% (by GC)
3-Chloro-1-fluoro-4-nitrobenzene
分子式:
C₆H₃ClFNO₂ 分子量: 175.55 g/mol 沸点: 71°C |
MDL编号:
MFCD03412200 CAS号: 2106-50-5 EINECS: 218-286-2 |
|
产品技术规格
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 35.0 to 39.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...