2,5-二氨基苯磺酸 ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 1,4-Phenylenediamine-2-sulphonic acid, 2,5-Diaminobenzenesulphonic acid

P1691-25G P1691-5G
TCIAP1691-25GEA 0 CNY
TCIAP1691-25G TCIAP1691-5G
2,5-二氨基苯磺酸 ≥98.0% (by HPLC, titration analysis)
2,5-二氨基苯磺酸
分子式: C₆H₈N₂O₃S
分子量: 188.21 g/mol
沸点: 500
溶点: 300°C
MDL编号: MFCD00007904
CAS号: 88-45-9
EINECS: 201-832-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed6bdec0502797a88b9742880b62f0d9324bba5f&vid=ed6bdec0502797a88b9742880b62f0d9324bba5f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light gray to Light orange powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR