N-苯基对苯二胺 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 对氨基二苯胺, 4-氨基二苯胺

P0203-500G P0203-25G
TCIAP0203-500GEA 0 CNY
TCIAP0203-500G TCIAP0203-25G
N-苯基对苯二胺 ≥98.0% (by HPLC, titration analysis)
N-苯基对苯二胺
分子式: C₁₂H₁₂N₂
分子量: 184.24 g/mol
沸点: 354°C
闪点: 193
MDL编号: MFCD00007850
CAS号: 101-54-2
EINECS: 202-951-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5de1156bf3b20cf706f4a6af0170d5e0e56d2122&vid=5de1156bf3b20cf706f4a6af0170d5e0e56d2122&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light gray to Brown to Black powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 73.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR