2-氯苯磺酰氯 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
2-Chlorobenzene-1-sulphonyl chloride
C2630-5G
C2630-25G
TCIAC2630-5GEA
0
CNY
TCIAC2630-5G
TCIAC2630-25G
2-氯苯磺酰氯 ≥98.0% (by GC, titration analysis)
2-氯苯磺酰氯
分子式:
C₆H₄Cl₂O₂S 分子量: 211.07 g/mol 沸点: 146°C 溶点: 28 密度: 1.548 闪点: 60°C |
MDL编号:
MFCD00051520 CAS号: 2905-23-9 EINECS: 220-807-3 UN号: 2920 ADR: 8 II |
|
产品技术规格
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...