缩二脲 ≥98.0% (以总氮计) (contains ca. 10% Triuret)

供应商: TCI
Warning

B0513-25G B0513-500G
TCIAB0513-25GEA 0 CNY
TCIAB0513-25G TCIAB0513-500G
缩二脲 ≥98.0% (以总氮计) (contains ca. 10% Triuret)
缩二脲
分子式: C₂H₅N₃O₂
溶点: 188 °C
密度: 1,467 g/cm³ (10 °C)
存储条件: Refrigerator
MDL编号: MFCD00007946
CAS号: 108-19-0
EINECS: 203-559-0
Merck Index: 14,01309
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acdcfa7715af871c05df5d9652cf72543032fb40&vid=acdcfa7715af871c05df5d9652cf72543032fb40&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Water almost transparency
Triuret max. 10.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR