5-氟-2-甲基苯甲酸 ≥98.0% (by GC)

供应商: TCI
Warning

F0844-5G F0844-25G
TCIAF0844-5GEA 0 CNY
TCIAF0844-5G TCIAF0844-25G
5-氟-2-甲基苯甲酸 ≥98.0% (by GC)
5-氟-2-甲基苯甲酸
分子式: C₈H₇FO₂
分子量: 154.14 g/mol
沸点: 263
MDL编号: MFCD00042294
CAS号: 33184-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=593a84b4a2855292014de40d504ffea753b3108f&vid=593a84b4a2855292014de40d504ffea753b3108f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR