邻甲苯磺酰氯 ≥75.0% (by GC) (contains ca. 23% isomer)
供应商: TCI
|
Danger
|
Synonyms:
Toluene-o-sulphonyl chloride, Toluene-2-sulphonyl chloride, 2-Toluenesulphonyl chloride
T0284-25G
T0284-500G
TCIAT0284-25GEA
0
CNY
TCIAT0284-25G
TCIAT0284-500G
邻甲苯磺酰氯 ≥75.0% (by GC) (contains ca. 23% isomer)
邻甲苯磺酰氯
分子式:
C₇H₇ClO₂S 分子量: 190.65 g/mol 沸点: 272 溶点: 10.2 |
MDL编号:
MFCD00024874 CAS号: 133-59-5 EINECS: 205-113-0 UN号: 3265 ADR: 8 III |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 75.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...