3,5-二苄氧基苯甲醛 ≥98.0% (by GC)

供应商: TCI

Synonyms: 3,5-Dibenzyloxybenzaldehyde

D3184-1G D3184-5G
TCIAD3184-1GEA 0 CNY
TCIAD3184-1G TCIAD3184-5G
3,5-二苄氧基苯甲醛 ≥98.0% (by GC)
3,5-二苄氧基苯甲醛
分子式: C₂₁H₁₈O₃
分子量: 318.3658 g/mol
MDL编号: MFCD00075777
CAS号: 14615-72-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fdde93daa9d83d07a071f28263d3dc0b9c921b4&vid=8fdde93daa9d83d07a071f28263d3dc0b9c921b4&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR