2-哌嗪酮 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: Piperazin-2-one

P1867-1G P1867-5G
TCIAP1867-1GEA 0 CNY
TCIAP1867-1G TCIAP1867-5G
2-哌嗪酮 ≥98.0% (by GC)
2-哌嗪酮
分子式: C₄H₈N₂O
沸点: 164°C
MDL编号: MFCD01318687
CAS号: 5625-67-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54b877030ecbb069c1421d1083370c3eb7f4507f&vid=54b877030ecbb069c1421d1083370c3eb7f4507f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 131.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR