4′-吗啉基苯乙酮 99%
供应商: Thermo Fisher Scientific
|
Warning
|
152900050
152900500
ACRO152900050EA
0
CNY
ACRO152900050
ACRO152900500
4′-吗啉基苯乙酮 99%
4′-吗啉基苯乙酮
分子式:
C₁₂H₁₅NO₂ 溶点: 94…98 °C 存储条件: Ambient |
MDL编号:
MFCD00006168 CAS号: 39910-98-0 EINECS: 254-698-9 |
产品技术规格
Appearance | Yellow to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 94 to 98 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...