2,3,4-三氟甲苯 ≥97.0% (by GC)

供应商: TCI
Warning

T2735-1G T2735-5G
TCIAT2735-1GEA 0 CNY
TCIAT2735-1G TCIAT2735-5G
2,3,4-三氟甲苯 ≥97.0% (by GC)
2,3,4-三氟甲苯
分子式: C₇H₅F₃
闪点: 28°C
MDL编号: MFCD00075194
CAS号: 193533-92-5
UN号: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e30a0c577435467dc1a83c9b55ee45e22f21aee5&vid=e30a0c577435467dc1a83c9b55ee45e22f21aee5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR