5-Fluoronicotinic acid ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

F0738-1G F0738-5G
TCIAF0738-1GEA 0 CNY
TCIAF0738-1G TCIAF0738-5G
5-Fluoronicotinic acid ≥98.0% (by GC, titration analysis)
5-Fluoronicotinic acid
分子式: C₆H₄FNO₂
分子量: 141.10 g/mol
MDL编号: MFCD01318537
CAS号: 402-66-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef791a56ab2740e766853e435af5c542d6c87b51&vid=ef791a56ab2740e766853e435af5c542d6c87b51&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR