5-氟-2-甲基苯甲醛 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
F0746-25G
F0746-5G
TCIAF0746-25GEA
0
CNY
TCIAF0746-25G
TCIAF0746-5G
5-氟-2-甲基苯甲醛 ≥95.0% (by GC)
5-氟-2-甲基苯甲醛
分子式:
C₈H₇FO 分子量: 138.14 g/mol 沸点: 99°C 闪点: 77°C |
MDL编号:
MFCD02261763 CAS号: 22062-53-9 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...