顺-13-二十二碳烯酸 ≥85.0% (通过GC)
供应商: TCI
D1008-25G
D1008-500G
TCIAD1008-25GEA
0
CNY
TCIAD1008-25G
TCIAD1008-500G
顺-13-二十二碳烯酸 ≥85.0% (通过GC)
顺-13-二十二碳烯酸
分子式:
C₂₂H₄₃NO |
MDL编号:
MFCD00882379 CAS号: 112-84-5 EINECS: 204-009-2 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 85.0 % |
Melting point | 79.0 to 85.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...