6-Bromo-o-cresol ≥94.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-Bromo-6-methylphenol

B3126-25G B3126-5G
TCIAB3126-25GEA 0 CNY
TCIAB3126-25G TCIAB3126-5G
6-Bromo-o-cresol ≥94.0% (by GC)
6-Bromo-o-cresol
分子式: C₇H₇BrO
分子量: 187.0300 g/mol
沸点: 208°C
溶点: 16°C
MDL编号: MFCD09907876
CAS号: 13319-71-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3d96205988738e7e722fde9e7a74c6a971a7c35&vid=c3d96205988738e7e722fde9e7a74c6a971a7c35&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR