3-Methyl-1-hexanol ≥97.0% (by GC)

供应商: TCI

M1246-1G M1246-5G
TCIAM1246-1GEA 0 CNY
TCIAM1246-1G TCIAM1246-5G
3-Methyl-1-hexanol ≥97.0% (by GC)
3-Methyl-1-hexanol
分子式: C₇H₁₆O
沸点: 96°C
MDL编号: MFCD01321178
CAS号: 13231-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86e1e7cfad45e3014dd65dce8820a8d3af4a155f&vid=86e1e7cfad45e3014dd65dce8820a8d3af4a155f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR