2,2′-二羟基二苯甲酮 ≥99.0% (通过GC)
供应商: TCI
|
Warning
|
D1099-10G
D1099-25G
TCIAD1099-10GEA
0
CNY
TCIAD1099-10G
TCIAD1099-25G
2,2′-二羟基二苯甲酮 ≥99.0% (通过GC)
2,2′-二羟基二苯甲酮
分子式:
C₁₃H₁₀O₃ |
MDL编号:
MFCD00002217 CAS号: 835-11-0 EINECS: 212-639-4 |
|
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystaline |
Purity (GC) | min. 99.0 % |
Melting point | 61.0 to 64.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...