2-Ethylhexyl diphenyl phosphate ≥90.0% (by GC)

供应商: TCI
Warning

P1021-25G P1021-500G
TCIAP1021-25GEA 0 CNY
TCIAP1021-25G TCIAP1021-500G
2-Ethylhexyl diphenyl phosphate ≥90.0% (by GC)
2-Ethylhexyl diphenyl phosphate
分子式: C₂₀H₂₇O₄P
分子量: 362.3998 g/mol
沸点: 232°C
溶点: –30°C
闪点: 224°C
MDL编号: MFCD00059949
CAS号: 1241-94-7
EINECS: 214-987-2
UN号: 3082
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83be00f3677658ceea3a13de6cf4dc2acd8d58e5&vid=83be00f3677658ceea3a13de6cf4dc2acd8d58e5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %
Specific gravity (20/20) 1.0870 to 1.0930
Refractive index n20/D 1.5080 to 1.5110


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR