溴代三(二甲基氨基)磷鎓六氟磷酸盐 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Danger

Synonyms: BroP

B3838-5G B3838-1G
TCIAB3838-5GEA 0 CNY
TCIAB3838-5G TCIAB3838-1G
溴代三(二甲基氨基)磷鎓六氟磷酸盐 ≥98.0% (by HPLC, titration analysis)
溴代三(二甲基氨基)磷鎓六氟磷酸盐
分子式: C₆H₁₈BrF₆N₃P₂
MDL编号: MFCD00191864
CAS号: 50296-37-2
UN号: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fee72b483f4b80aa858254eed6734be10a9b729d&vid=fee72b483f4b80aa858254eed6734be10a9b729d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR