4′-(甲磺酰)苯乙酮 ≥98.0% (by GC)
供应商: TCI
Synonyms:
4-(甲磺酰)苯乙酮
M1678-25G
M1678-250G
TCIAM1678-25GEA
0
CNY
TCIAM1678-25G
TCIAM1678-250G
4′-(甲磺酰)苯乙酮 ≥98.0% (by GC)
4′-(甲磺酰)苯乙酮
分子式:
C₉H₁₀O₃S 分子量: 198.24 g/mol |
MDL编号:
MFCD00025072 CAS号: 10297-73-1 EINECS: 233-672-0 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 127.0 to 131.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...