p-Toluenesulphonylacetonitrile ≥99.0% (by GC)

供应商: TCI
Danger

Synonyms: [(4-Methylphenyl)sulphonyl]acetonitrile

T0846-25G T0846-5G
TCIAT0846-25GEA 0 CNY
TCIAT0846-25G TCIAT0846-5G
p-Toluenesulphonylacetonitrile ≥99.0% (by GC)
p-Toluenesulphonylacetonitrile
分子式: C₉H₉NO₂S
MDL编号: MFCD00039487
CAS号: 5697-44-9
EINECS: 227-173-7
UN号: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5025876e55497bc175b619cac5e33e3d6005c6c9&vid=5025876e55497bc175b619cac5e33e3d6005c6c9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 149.0 to 152.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR