2-Amino-5-chloro-4-methylbenzenesulphonic acid ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

A0287-25G A0287-500G
TCIAA0287-25GEA 0 CNY
TCIAA0287-25G TCIAA0287-500G
2-Amino-5-chloro-4-methylbenzenesulphonic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-5-chloro-4-methylbenzenesulphonic acid
分子式: C₇H₈ClNO₃S
MDL编号: MFCD00043924
CAS号: 88-53-9
EINECS: 201-839-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07813d50bb0164bef685aa54fbcf4e5616c52fe9&vid=07813d50bb0164bef685aa54fbcf4e5616c52fe9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot NaOH very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR