2-Bromo-4'-(methylsulphonyl)acetophenone ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 2-Bromo-1-(4-(methylsulphonyl)phenyl)ethanone, 4-(Methylsulphonyl)phenacyl bromide

B2573-25G B2573-5G
TCIAB2573-25GEA 0 CNY
TCIAB2573-25G TCIAB2573-5G
2-Bromo-4'-(methylsulphonyl)acetophenone ≥98.0% (by GC)
2-Bromo-4'-(methylsulphonyl)acetophenone
分子式: C₉H₉BrO₃S
分子量: 277.1350 g/mol
MDL编号: MFCD00673134
CAS号: 50413-24-6
UN号: 1759
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c8d920b27b7ced6f8dcefc4942657fae0e1d17e&vid=5c8d920b27b7ced6f8dcefc4942657fae0e1d17e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR