4-Methoxydiphenylamine ≥98.0% (通过GC)

供应商: TCI
Warning

M2069-25G M2069-5G
TCIAM2069-25GEA 0 CNY
TCIAM2069-25G TCIAM2069-5G
4-Methoxydiphenylamine ≥98.0% (通过GC)
4-Methoxydiphenylamine
分子式: C₁₃H₁₃NO
沸点: 195°C
MDL编号: MFCD00228649
CAS号: 1208-86-2
EINECS: 214-902-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=042e8343e60c962eb39e8f48bf38f091b2587f7e&vid=042e8343e60c962eb39e8f48bf38f091b2587f7e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR