4-Fluorodiphenylamine ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4-Fluoro-N-phenylaniline
F0630-1G
F0630-5G
TCIAF0630-1GEA
0
CNY
TCIAF0630-1G
TCIAF0630-5G
4-Fluorodiphenylamine ≥98.0% (by GC)
4-Fluorodiphenylamine
分子式:
C₁₂H₁₀FN 分子量: 187.2129 g/mol 沸点: 166°C 存储条件: Refrigerator |
MDL编号:
MFCD01318538 CAS号: 330-83-6 |
|
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 36.0 to 40.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...