5-Pyrimidineboronic acid (contains varying amounts of Anhydride)
供应商: TCI
|
Warning
|
Synonyms:
Pyrimidine-5-boronic acid, 5-Pyrimidylboronic acid, Pyrimidin-5-ylboronic acid, 5-Pyrimidinylboronic acid
P1759-1G
P1759-100MG
TCIAP1759-1GEA
0
CNY
TCIAP1759-1G
TCIAP1759-100MG
5-Pyrimidineboronic acid (contains varying amounts of Anhydride)
5-Pyrimidineboronic acid
分子式:
C₄H₅BN₂O₂ 存储条件: Refrigerator |
MDL编号:
MFCD03002366 CAS号: 109299-78-7 |
|
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | 95.0 to 117.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...