4-碘-1-甲基-1H-吡唑 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
I0829-1G
I0829-5G
TCIAI0829-1GEA
0
CNY
TCIAI0829-1G
TCIAI0829-5G
4-碘-1-甲基-1H-吡唑 ≥98.0% (by GC)
4-碘-1-甲基-1H-吡唑
分子式:
C₄H₅IN₂ |
MDL编号:
MFCD05663860 CAS号: 39806-90-1 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 62.0 to 66.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...