4-溴-3-甲基-1H-吡唑 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 4-Bromo-3-methylpyrazole

B3522-1G B3522-5G
TCIAB3522-1GEA 0 CNY
TCIAB3522-1G TCIAB3522-5G
4-溴-3-甲基-1H-吡唑 ≥98.0% (by GC)
4-溴-3-甲基-1H-吡唑
分子式: C₄H₅BrN₂
分子量: 161.01 g/mol
MDL编号: MFCD00005241
CAS号: 13808-64-5
EINECS: 237-456-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6771f93f780b451ceabaad57ef829f24cd65a992&vid=6771f93f780b451ceabaad57ef829f24cd65a992&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 74.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR