2,3-二甲基马来酸酐 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 二甲基马来酸酐

116390050 116390500
ACRO116390050EA 0 CNY
ACRO116390050 ACRO116390500
2,3-二甲基马来酸酐 97%
2,3-二甲基马来酸酐
分子式: C₆H₆O₃
分子量: 126.11 g/mol
溶点: 91…96 °C
存储条件: Ambient
MDL编号: MFCD00005523
CAS号: 766-39-2
EINECS: 212-165-8

现在订购

产品技术规格

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 91 to 96 °C
Titration with TBAH ≥96.0 %
Chloride (Cl) ≤5 ppm
Nitrite (NO2) (Limit about 5 ppm) Passes test
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.002 %
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR