N-邻苯二甲酰甘氨酸 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
邻苯二甲酰甘氨酸, 1,3-二氧-2-异吲哚啉乙酸
P0963-25G
P0963-500G
TCIAP0963-25GEA
0
CNY
TCIAP0963-25G
TCIAP0963-500G
N-邻苯二甲酰甘氨酸 ≥98.0% (by HPLC, titration analysis)
N-邻苯二甲酰甘氨酸
分子式:
C₁₀H₇NO₄ |
MDL编号:
MFCD00005900 CAS号: 4702-13-0 EINECS: 225-177-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...