双联频哪醇硼酸酯

供应商: LOCAL DEALS

Synonyms: 双戊酰二硼

R01071810124 R01071810111
CHLCR01071810124EA 0 CNY
CHLCR01071810124 CHLCR01071810111
双联频哪醇硼酸酯
双联频哪醇硼酸酯
分子式: C₁₂H₂₄B₂O₄
分子量: 253.94 g/mol
MDL编号: MFCD00799570
CAS号: 73183-34-3

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR