[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex

供应商: LOCAL DEALS

R02178680131 R02178680144
CHLCR02178680131EA 0 CNY
CHLCR02178680131 CHLCR02178680144
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
[1,1′-双(二苯基膦)二茂铁]二氯化钯(II)二氯甲烷络合物,ChemDose,[1,1′-双(二苯基膦)二茂铁]二氯化钯(II)二氯甲烷络合物浸渍片剂
分子式: C₃₅H₃₀Cl₄FeP₂Pd
MDL编号: MFCD00792899
CAS号: 95464-05-4

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR