[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride

供应商: LOCAL DEALS

R00287080121 R00287080144 R00287080133
CHLCR00287080121EA 0 CNY
CHLCR00287080121 CHLCR00287080144 CHLCR00287080133
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride
[1,1′-双(二苯基膦)二茂铁]二氯化钯(II)
分子式: C₃₄H₂₈Cl₂P₂FePd
MDL编号: MFCD00015757
CAS号: 72287-26-4
EINECS: 460-040-3

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR