[1,1′-双(二苯基膦)二茂铁]二氯化钴(II) ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]cobalt(II)
B3374-1G
B3374-5G
TCIAB3374-1GEA
0
CNY
TCIAB3374-1G
TCIAB3374-5G
[1,1′-双(二苯基膦)二茂铁]二氯化钴(II) ≥98.0% (by titrimetric analysis)
[1,1′-双(二苯基膦)二茂铁]二氯化钴(II)
分子式:
C₃₄H₂₈Cl₂CoFeP₂ 分子量: 684.23 g/mol |
MDL编号:
MFCD03427161 CAS号: 67292-36-8 |
|
产品技术规格
Appearance | Yellow to Brown to Dark green powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...