甲基乙烯基砜 95% stabilised
供应商: Thermo Fisher Scientific
|
Danger
|
306730100
306730010
ACRO306730100EA
0
CNY
ACRO306730100
ACRO306730010
甲基乙烯基砜 95% stabilised
甲基乙烯基砜
分子式:
C₃H₆O₂S 分子量: 106.15 g/mol 沸点: 115…117 °C (19 mmHg) 密度: 1.21 g/cm³ 闪点: >110 °C 存储条件: Refrigerator |
MDL编号:
MFCD00007567 CAS号: 3680-02-2 EINECS: 222-958-0 UN号: 2810 ADR: 6.1 I |
产品技术规格
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4610 to 1.4650 (20°C, 589 nm) |
GC | ≥94 % |
Stabilizer | 200 ppm 4-tert-Butylphenol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...