3-氨基丙腈 99+%
供应商: Thermo Fisher Scientific
|
Warning
|
104470250
104471000
104470050
ACRO104470250EA
0
CNY
ACRO104470250
ACRO104471000
ACRO104470050
3-氨基丙腈 99+%
3-氨基丙腈
分子式:
C₁₀H₁₆N₄O₄ 分子量: 256.26 g/mol 溶点: 170…173 °C (dec.) 存储条件: Ambient |
MDL编号:
MFCD00013142 CAS号: 2079-89-2 EINECS: 218-208-7 |
产品技术规格
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥99.0 % |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...