4,7,10-三氧-1,13-十三烷二胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: O,O′-双(3-氨丙基)二乙二醇

D1571-25ML D1571-500ML
TCIAD1571-25MLEA 0 CNY
TCIAD1571-25ML TCIAD1571-500ML
4,7,10-三氧-1,13-十三烷二胺 ≥98.0% (by GC, titration analysis)
4,7,10-三氧-1,13-十三烷二胺
分子式: C₁₀H₂₄N₂O₃
分子量: 220.3092 g/mol
沸点: 151°C
闪点: 166°C
MDL编号: MFCD00059850
CAS号: 4246-51-9
EINECS: 224-207-2
UN号: 2735
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e9bd840960b10e8fd002c56c9cfa4ece35bf51f&vid=8e9bd840960b10e8fd002c56c9cfa4ece35bf51f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR