Electroporation cuvettes, Gene Pulser Xcell™
供应商: BIO RAD
165-2086 165-2088 165-2091 165-2092 165-2093
BRAD165-2086EA 0 CNY
BRAD165-2086 BRAD165-2088 BRAD165-2091 BRAD165-2092 BRAD165-2093
Electroporation cuvettes, Gene Pulser Xcell™
比色皿 电击杯
Gene Pulser®/Micropulser™ electroporation cuvettes are high-quality electroporation cuvettes which enable reproducible and consistent gene delivery to valuable transfection samples.
  • Universal electroporation systems
  • Wash thoroughly to eliminate trace inhibitor materials
  • Sterilised and individually wrapped
  • Reliable sturdy, seamless construction with gap widths set to exacting tolerances for consistent performance
Multiple configurations available in three gap widths and sample type: 0,4, 0,2, and 0,1 cm to deliver the proper field strength to a variety of cell types:
0,1 cm (brown cap) - E. coli, bacteria, yeast
0,2 cm (green cap) - E. coli, bacteria, yeast, mammalian cells, including primary and stem cells
0,4 cm (blue cap) - Mammalian cells, including primary and stem cells, and plant protoplasts