Precast ExcelGel for SDS-PAGE
供应商: GE HEALTHCARE
17-1342-01 80-1255-53
GEHE17-1342-01EA 0 CNY
GEHE17-1342-01 GEHE80-1255-53
Precast ExcelGel for SDS-PAGE
凝胶电泳 预制凝胶
ExcelGel precast gels for SDS-PAGE are available as homogeneous or gradient gels. The homogeneous gels have 25 preformed sample wells for sample volumes from 5 to 17 µl. The gradient gels use sample application strips (26 samples up to 40 µl, 52 samples up to 20 µl), paper pieces or sample droplets. All ExcelGel SDS homogeneous and 8 - 18 gradient gels have a 33 mm stacking zone and a 77 mm separating zone.
  • Save time with ready-to-use, mechanically stable gels cast on plastic supports
  • Apply samples quickly and easily using preformed sample wells
  • Avoid