(2-Chloro-4-pyridinyl)methanol

供应商: LOCAL DEALS

R00839820131 R00839820133
CHLCR00839820131EA 0 CNY
CHLCR00839820131 CHLCR00839820133
(2-Chloro-4-pyridinyl)methanol
(2-氯-4-吡啶基)甲醇
分子式: C₆H₆ClNO
MDL编号: MFCD06858767
CAS号: 100704-10-7

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR