吡啶

供应商: LOCAL DEALS
Danger

Synonyms: Azabenzene, Azine

R00268620024 R00268620024CD
CHLCR00268620024EA 0 CNY
CHLCR00268620024 CHLCR00268620024CD
吡啶
吡啶
分子式: C₅H₅N
分子量: 79.1 g/mol
沸点: 115
溶点: –44 °C
密度: 0.983 g/cm³ (20 °C)
闪点: 17
存储条件: Ambient
MDL编号: MFCD00011732
CAS号: 110-86-1
EINECS: 203-809-9
UN号: 1282
ADR: 3 II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR