氯化铝

供应商: LOCAL DEALS
Danger

Synonyms: Aluminum chloride

R02140720124 R02140720124CD
CHLCR02140720124EA 0 CNY
CHLCR02140720124 CHLCR02140720124CD
氯化铝
氯化铝
分子式: AlCl₃
沸点: 180 °C (1013 hPa)
溶点: 180…181 °C
密度: liquid 1,31 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00003422
CAS号: 7446-70-0
EINECS: 231-208-1
UN号: 1726
ADR: 8,II
Merck Index: 13,00335

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR