碳酸钾

供应商: LOCAL DEALS
Warning

R01273920024CD R01273920024
CHLCR01273920024CDEA 0 CNY
CHLCR01273920024CD CHLCR01273920024
碳酸钾
碳酸钾
分子式: K₂CO₃
溶点: 891 °C
密度: 2,42 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00011382
CAS号: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR