喹哪啶酸

供应商: LOCAL DEALS
Warning

Synonyms: 2-喹啉羧酸

R01845110033 R01845110121
CHLCR01845110033EA 0 CNY
CHLCR01845110033 CHLCR01845110121
喹哪啶酸
喹哪啶酸
分子式: C₁₀H₇NO₂
分子量: 173.17 g/mol
溶点: 154…157 °C
存储条件: Ambient
MDL编号: MFCD00006752
CAS号: 93-10-7
EINECS: 202-218-3
Merck Index: 12,08227

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR