2'-Trifluoromethoxyacetophenone ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-[2-(Trifluoromethoxy)phenyl]-1-ethanone, 1-(2-(Trifluoromethoxy)phenyl)ethanone
T2794-5G
T2794-25G
TCIAT2794-5GEA
0
CNY
TCIAT2794-5G
TCIAT2794-25G
2'-Trifluoromethoxyacetophenone ≥98.0% (by GC)
2'-Trifluoromethoxyacetophenone
分子式:
C₉H₇F₃O₂ 分子量: 204.15 |
MDL编号:
MFCD00236320 CAS号: 220227-93-0 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...