乙二醇

供应商: LOCAL DEALS
Warning

R00264720016 R00264720024
CHLCR00264720016EA 0 CNY
CHLCR00264720016 CHLCR00264720024
乙二醇
乙二醇
分子式: HOCH₂CH₂OH
沸点: 196…198 °C (1013 hPa)
溶点: –13 °C
密度: 1,115 g/cm³ (20 °C)
闪点: 116 °C
存储条件: Refrigerator
MDL编号: MFCD00002885
CAS号: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR