2-Propanol

供应商: LOCAL DEALS
Danger

Synonyms: 仲丙醇, 异丙基醇, 异丙醇, IPA

R01271920024 R01271920024CD
CHLCR01271920024EA 0 CNY
CHLCR01271920024 CHLCR01271920024CD
2-Propanol
2-丙醇
分子式: (CH₃)₂CHOH
分子量: 60.1 g/mol
沸点: 82
溶点: –89 °C
密度: 0.786 g/cm³ (20 °C)
闪点: 12 °C
存储条件: Ambient
MDL编号: MFCD00011674
CAS号: 67-63-0
EINECS: 200-661-7
UN号: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR