乙酸 glacial

供应商: LOCAL DEALS
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

R00288620024CD R00288620116 R00288620024 R00288620124
CHLCR00288620024CDEA 0 CNY
CHLCR00288620024CD CHLCR00288620116 CHLCR00288620024 CHLCR00288620124
乙酸 glacial
乙酸
分子式: H₃CCOOH
分子量: 60.05 g/mol
沸点: 118
溶点: 17
密度: 1.05 g/cm³ (20 °C)
闪点: 38.5
存储条件: Ambient
MDL编号: MFCD00036152
CAS号: 64-19-7
EINECS: 200-580-7
UN号: 2789
ADR: 8 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR