乙酸乙酯

供应商: LOCAL DEALS
Danger

Synonyms: Acetic acid ethyl ester

R00280320024CD R00280320024
CHLCR00280320024CDEA 0 CNY
CHLCR00280320024CD CHLCR00280320024
乙酸乙酯
乙酸乙酯
分子式: CH₃COOC₂H₅
分子量: 88.11 g/mol
沸点: 77.1
溶点: –83 °C
密度: 0.902 g/cm³ (20 °C)
闪点: –4 °C
存储条件: Ambient
MDL编号: MFCD00009171
CAS号: 141-78-6
EINECS: 205-500-4
UN号: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR